Learn More
Applied Biosystems™ PureQuant™ Treg Assay
Shop All Applied Biosystems ProductsDescription
Assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples. Results are provided as a percentage of target cell type out of total input cells using a free analysis tool.
The PureQuant™ Treg Assay utilizes a specific demethylated site in the transcription factor FOXP3 gene to positively identify regulatory T-cells. This specific site allows for discrimination of regulatory T-cells from activated CD4+ FOXP3+ conventional T-cells
Benefits include:
- Proven performance―core technology has been used for identification and quantification of immune cell types for over a decade
- Easily standardized―qPCR method enables consistent results across multiple users, instruments, and sites
- Facilitates regulatory filings―regulatory support file provided upon request; established performance specifications following ICHQ2(R1) guidelines
- Scheduling flexibility―assay can be performed using fresh or frozen cells/gDNA
Assays include all reagents, standards, and controls required to run the assay. Specific components include:
- Calibrator: Plasmid-based calibrator harbors the genomic target regions in equimolar ratios and includes GAPDH. Subjected to bisulfite conversion followed by copy number estimation via qPCR standard curve. Allows estimation of assay-specific bisulfite efficiency factor: the ratio between TpG/GAPDH copy numbers measured from the same plasmid.
- Reference: checks assay performance. Consists of genomic DNA extracted from human blood of pooled healthy donors. The reference values represent the expected range of the target cell type. For example, CD8+ cells are generally more abundant than Treg or Th17 cells.
- Standards: determine the unknown copy number. Consist of six dilutions spanning 4-log linear dynamic range (30-31250 copies). Absolute copy numbers can reliably be determined using the slope and intercept of the respective standard curves.
This product is manufactured at a site with an ISO9001-certified quality management system.
Specifications
Specifications
| Content And Storage |
|
| Detection Method | Primer-Probe Detection |
| For Use With (Application) | Cells, DNA |
| Quantity | 28 reactions |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.