Learn More
Sartorius™ Cubis™ II High-Capacity Lab Balance
With a maximum load between 11.2 and 70.2 kilograms and a readability between 0.1 and 1 gram, this balance provides the ideal model for every application.
Brand: Sartorius™ MCA36201S-2CEU-0
Description
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Sartorius™ Cubis™ II balance series.
- All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems
- Design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement
- Modern user interfaces and easy process integration
- Pharmaceutical and GxP compliance
- Data handling, integrity and connectivity
- Ergonomic sample handling
- Unlimited communication at the highest level of accuracy and precision
Specifications
Type Approval Certificate for China, CE | |
36,200 g | |
100 mg | |
1.5 s | |
100/240 VAC (±10%), 50/60 Hz, 1 A |
D-sub Connector, 9-pin | |
Color Touchscreen | |
Aluminum, PBT, Optiwhite Float Glass, Stainless Steel | |
Electronic Toploading Balance | |
No |