Learn More
Thermo Scientific™ Culti-Loops™ API™ Coryne QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4653063
Product Code. 15219277
  • 610.00 EUR / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Description
Thermo Scientific™ Culti-Loops™ API Coryne Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Cellulosimicrobium cellulans ATCC™ 27402™†
Corynebacterium renale ATCC™ 19412™†
Microbacterium testaceum ATCC™ 15829™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
27402TM*
Culti-Loops
Organism Sets
4 Loops/Pack
API Coryne QC Set/3
Culti-Loops QC Set
Qualitative QC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.