Learn More
Thermo Scientific™ Culti-Loops™ API™ NH QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4653070
Product Code. 15420871
  • 610.00 EUR / Pack of 15
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Description
Thermo Scientific™ Culti-Loops™ API NH Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Haemophilus influenzae Type b ATCC™ 10211™†
Haemophilus paraphrophilus ATCC™ 49917™†
Neisseria gonorrhoeae ATCC™ 31426™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10211TM*
Culti-Loops
Organism Sets
1 Loop/Pack
API NH QC Set/3
Culti-Loops QC Set
Qualitative QC
1/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.