Learn More
Tocris Bioscience™ CX 614
AMPAkine; positive allosteric modulator of AMPA
Brand:  Tocris Bioscience™ 5149/50
723.95 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18716939
  • 915.00 EUR / 50mg
Explore more special offers
Description
Description
CX 614 is an AMPAkine; positive allosteric modulator of AMPA. Prototypical benzamide which occupies subsite A only. Modulates channel deactivation and desensitization.
Specifications
Specifications
>99%
CX 614
CX 614
50 mg
247.25