missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cycloheptyl bromide, 97%, Thermo Scientific Chemicals
91.35 EUR - 350.75 EUR
Chemical Identifiers
CAS | 2404-35-5 |
---|---|
Molecular Formula | C7H13Br |
Molecular Weight (g/mol) | 177.09 |
MDL Number | MFCD00004149 |
InChI Key | LOXORFRCPXUORP-UHFFFAOYSA-N |
Synonym | cycloheptane, bromo, cycloheptyl bromide, cycloheptylbromide, bromocyclo-heptane, 2-bromocycloheptane, bromocycloheptane, acmc-209g8c |
PubChem CID | 16992 |
IUPAC Name | bromocycloheptane |
SMILES | BrC1CCCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10606682
|
Thermo Scientific Acros
111080250 |
25 g | Glass bottle |
91.35 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10616682
|
Thermo Scientific Acros
111081000 |
100 g | Glass bottle |
350.75 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2404-35-5 | |
177.09 | |
LOXORFRCPXUORP-UHFFFAOYSA-N | |
16992 | |
BrC1CCCCCC1 |
C7H13Br | |
MFCD00004149 | |
cycloheptane, bromo, cycloheptyl bromide, cycloheptylbromide, bromocyclo-heptane, 2-bromocycloheptane, bromocycloheptane, acmc-209g8c | |
bromocycloheptane |
Specifications
2404-35-5 | |
1.2800g/mL | |
68°C | |
96% min. (GC) | |
C7H13Br | |
MFCD00004149 | |
1.28 | |
LOXORFRCPXUORP-UHFFFAOYSA-N | |
bromocycloheptane | |
16992 | |
97% | |
Cycloheptyl bromide, 97% |
Yellow | |
71°C to 72°C (10.0 mmHg) | |
Authentic | |
C7H13Br | |
1.5035 to 1.5055 | |
05, 29 | |
cycloheptane, bromo, cycloheptyl bromide, cycloheptylbromide, bromocyclo-heptane, 2-bromocycloheptane, bromocycloheptane, acmc-209g8c | |
BrC1CCCCCC1 | |
177.09 | |
177.08 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 219-294-9
missing translation for 'tsca' : TSCA
Product Identifier
- Cycloheptyl bromide
RUO – Research Use Only