missing translation for 'onlineSavingsMsg'
Learn More

cis-Cyclooctene, 95%, stabilized, Thermo Scientific Chemicals

71.66 EUR - 439.19 EUR

Chemical Identifiers

CAS 931-87-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00001753
InChI Key URYYVOIYTNXXBN-UPHRSURJSA-N
Synonym cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
PubChem CID 638079
SMILES C1CCC\C=C/CC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10011800
View Documents
Thermo Scientific Acros
154862500
250 mL Glass bottle
71.66 EUR
250mL
Please to purchase this item. Need a web account? Register with us today!
10255530
View Documents
Thermo Scientific Acros
154860010
1 L Glass bottle
204.02 EUR
1 L
Please to purchase this item. Need a web account? Register with us today!
11974431
View Documents
Thermo Scientific Acros
154860025
2.5 L Glass bottle
439.19 EUR
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

931-87-3
110.20
URYYVOIYTNXXBN-UPHRSURJSA-N
638079
C8H14
MFCD00001753
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
C1CCC\C=C/CC1

Specifications

931-87-3
Colorless to Brown
145.0°C to 146.0°C
Authentic
C8H14
1.4680 to 1.4710
0.84
Solubility in water: <0.1g/L (20°C)
C1CCC\C=C/CC1
100 to 200ppm irganox 1076 FD
638079
110.2
Liquid
-16.0°C
0.8400g/mL
25°C
94% min. (GC)
C8H14
MFCD00001753
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
URYYVOIYTNXXBN-UPHRSURJSA-N
cyclooctene
110.20
1.89 mPa.s (20°C)
95%
cis-Cyclooctene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Store in a well

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 213-243-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.