Learn More
Cyclopentanecarboxaldehyde, 97%, stabilized, Thermo Scientific Chemicals
47.84 EUR - 537.68 EUR
Chemical Identifiers
CAS | 872-53-7 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.14 |
InChI Key | VELDYOPRLMJFIK-UHFFFAOYSA-N |
Synonym | cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde |
PubChem CID | 70106 |
IUPAC Name | cyclopentanecarbaldehyde |
SMILES | C1CCC(C1)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10663985
|
Thermo Scientific Acros
425910010 |
1 g | Glass bottle |
47.84 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10356644
|
Thermo Scientific Acros
425910050 |
5 g | Glass bottle |
155.78 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964781
|
Thermo Scientific Acros
425910250 |
25 g | Glass bottle |
537.68 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
872-53-7 | |
98.14 | |
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde | |
cyclopentanecarbaldehyde |
C6H10O | |
VELDYOPRLMJFIK-UHFFFAOYSA-N | |
70106 | |
C1CCC(C1)C=O |
Specifications
872-53-7 | |
0.9190g/mL | |
28°C | |
96% min. (GC) | |
C5H9CHO | |
0.919 | |
VELDYOPRLMJFIK-UHFFFAOYSA-N | |
cyclopentanecarbaldehyde | |
98.14 | |
98.14 | |
Liquid |
Colorless to Yellow | |
140.0°C to 141.0°C | |
Authentic | |
C6H10O | |
1.4420 to 1.4460 (20°C, 589nm) | |
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde | |
C1CCC(C1)C=O | |
0.1 wt% hydroquinone | |
70106 | |
97% | |
Cyclopentanecarboxaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-829-7
- Cyclopentanecarboxaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only