Learn More
Cyclopropylacetylene, 97%
Product Code. 15400258
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 15400258
Brand: Thermo Scientific Alfa Aesar H53440.22
Request Bulk or Custom Format
CAS: 6746-94-7 | C5H6 | 66.103 g/mol
Cyclopropylacetylene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopropylacetylene is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6746-94-7
Molecular Formula C5H6
Molecular Weight (g/mol) 66.103
MDL Number MFCD02181090
InChI Key NPTDXPDGUHAFKC-UHFFFAOYSA-N
Synonym cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene
PubChem CID 138823
IUPAC Name ethynylcyclopropane
SMILES C#CC1CC1
Specifications
Density 0.781
Boiling Point 51°C
Flash Point −17°C (−1°F)
Refractive Index 1.429
Quantity 100 g
UN Number UN3295
Solubility Information Slightly soluble in water.
Formula Weight 66.1
Percent Purity 97%
Chemical Name or Material Cyclopropylacetylene