missing translation for 'onlineSavingsMsg'
Learn More

(Cyclopropylmethyl)triphenylphosphonium bromide, 98+%

Product Code. 11384474
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11384474

Brand: Thermo Scientific Alfa Aesar L01145.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14799-82-7 | C22H22BrP | 397.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14799-82-7
Molecular Formula C22H22BrP
Molecular Weight (g/mol) 397.30
MDL Number MFCD00051878
InChI Key WFQSHRSBITUSIB-UHFFFAOYSA-M
Synonym cyclopropylmethyl triphenylphosphonium bromide, cyclopropylmethyl triphenyl phosphanium bromide, cyclopropylmethyl triphenyl phosphonium bromide, phosphonium, cyclopropylmethyl triphenyl-, bromide, cyclopropylmethyl triphenylphosphanium bromide, acmc-1cg9v, cyclopropylmethyltriphenylphosphonium bromide, cyclopropylmethyl triphenylphosphoniumbromide, phosphonium, cyclopropylmethyl triphenyl-, bromide 1:1
PubChem CID 3084208
IUPAC Name cyclopropylmethyl(triphenyl)phosphanium;bromide
SMILES [Br-].C(C1CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 178°C to 182°C
Quantity 25 g
Beilstein 4068302
Sensitivity Hygroscopic
Formula Weight 397.29
Percent Purity ≥98%
Chemical Name or Material (Cyclopropylmethyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.