Learn More
D-Alanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
116.58 EUR - 397.98 EUR
Chemical Identifiers
CAS | 59531-86-1 |
---|---|
Molecular Formula | C7H16ClNO2 |
Molecular Weight (g/mol) | 181.66 |
MDL Number | MFCD00077087 |
InChI Key | WIQIWPPQGWGVHD-NUBCRITNSA-N |
Synonym | d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochlorideShow More |
PubChem CID | 16211919 |
IUPAC Name | tert-butyl (2R)-2-aminopropanoate;hydrochloride |
SMILES | CC(C(=O)OC(C)(C)C)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15476338
|
Thermo Scientific Alfa Aesar
H59898.03 |
1 g |
116.58 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15486338
|
Thermo Scientific Alfa Aesar
H59898.06 |
5 g |
397.98 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59531-86-1 | |
181.66 | |
WIQIWPPQGWGVHD-NUBCRITNSA-N | |
16211919 | |
CC(C(=O)OC(C)(C)C)N.Cl |
C7H16ClNO2 | |
MFCD00077087 | |
d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochlorideShow More | |
tert-butyl (2R)-2-aminopropanoate;hydrochloride |
Specifications
59531-86-1 | |
MFCD00077087 | |
d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride | |
CC(C(=O)OC(C)(C)C)N.Cl | |
181.66 | |
181.66 | |
Hygroscopic | |
D-Alanine tert-butyl ester hydrochloride |
C7H16ClNO2 | |
5763077 | |
WIQIWPPQGWGVHD-NUBCRITNSA-N | |
tert-butyl (2R)-2-aminopropanoate;hydrochloride | |
16211919 | |
−1.4° (c=2 in Ethanol) | |
98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.