missing translation for 'onlineSavingsMsg'
Learn More

D-Biotin, MP Biomedicals

Product Code. 11388599
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

  • D-Biotin may be used to elute proteins from avidin/streptavidin resins
  • It is widely used for dietary supplements and fortified foods
  • It is also used for tablets and hard-shell capsule preparation due to its pharmaceutical properties

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Melting Point 223°C to 233°C
Color White
Boiling Point 573.6°C (760 mmHg(literature))
Linear Formula C10H16N2O3S
Quantity 100 mg
Solubility Information Soluble in water (22 mg/100 mL), ethanol (80 mg/100 mL), more soluble in hot water and in dilute alkalies; insoluble in other common organic solvents. Soluble in 2 M Ammonium hydroxide (50mg/mL - clear, colorless solution), dimethylformamide (1.7mg/mL).
Formula Weight 244.3
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material D-BIOTIN
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'