missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Biotin, MP Biomedicals™
Click to view available options
Quantity:
500 mg
Description
D-biotin occurs mainly bound to proteins or polypeptides. It is more abundant in the liver, kidney, pancreas, yeast and milk. Biotin levels are higher in cancerous tumors than in normal tissues. It is inactivated by binding to avidin.
- Product is an essential vitamin that is important for amino acid and energy metabolism, and fatty acid synthesis.
- It is a prosthetic group in four mammalian carboxylase families and facilitates the binding and transfer of carbon dioxide.
Specifications
Specifications
CAS | 58-85-5 |
Melting Point | 223°C to 233°C |
Density | 1.267 g/cm3(Lit.) |
Boiling Point | 573.6°C (760 mmHg) |
Molecular Formula | C10H16N2O3S |
Refractive Index | 90.5° (C=2, 0.1mol/L NaOH)(Lit.) |
MDL Number | MFCD00005541 |
Quantity | 500 mg |
Synonym | biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin |
Solubility Information | Soluble in water (22 mg/100 mL), ethanol (80 mg/100 mL), more soluble in hot water and in dilute alkalies; insoluble in other common organic solvents. Soluble in 2 M Ammonium hydroxide (50mg/mL; clear, colorless solution), dimethylformamide (1.7 mg/mL). |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'