missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%

Product Code. p-4509598 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
10g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10122770

Brand: Thermo Scientific Chemicals 119900100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H13NO5·HCl

Specifications

Color Brown-Yellow to Yellow
Loss on Drying 1.0 % max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Quantity 10 g
Specific Rotation Condition + 72.50 (20.00°C c=2,H2O,5hrs.)
Specific Rotation +70.0° to +73.0° (20°C, 589nm) (c=2.5, H2O) (after 3 hrs)
Formula Weight 215.64
Percent Purity 98.0 to 102.0% (on dry substance) (Total base)
Physical Form Powder
Chemical Name or Material D-Glucosamine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.