missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD D- Luciferin Potassium Salt
Synthetically made substrate which can be catalyzed by firefly luciferase
725.00 EUR - 1900.00 EUR
Specifications
Formulation | Lyophilized |
---|---|
Type | D-Luciferin |
Content And Storage | -20°C |
For Use With (Application) | Luminometer |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15878788
|
BD
556879 |
100 mg | N/A | ||||||
15858788
|
BD
556877 |
10 mg | N/A | ||||||
15868788
|
BD
556878 |
50 mg |
725.00 EUR
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
15888788
|
BD
556888 |
250 mg |
1375.00 EUR
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
Description
- Also known as beetle luciferin
- Lyophilized in form of monopotassium salt
- Purified by HPLC to remove dehydroluciferin and L-Luciferin
- Form dehydroluciferin in presence of ultraviolet light and/or moisture
Luminometer
Specifications
Lyophilized | |
D-Luciferin | |
-20°C | |
Luminometer |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title