missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-ribulose, 99.7%, For HPLC analysis, MP Biomedicals™
Click to view available options
Quantity:
10 mg
25 mg
100 mg
500 mg
Specifications
Specifications
Color | Colorless |
Quantity | 10 mg |
Formula Weight | 150.1 |
Percent Purity | 0.997 |
Physical Form | Liquid |
Chemical Name or Material | D-ribulose, For HPLC analysis |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'