Learn More
Memmert™ DAkkS Factory Calibration for IF260plus
Product Code. 17506855 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17506855
Brand: Memmert™ IF260plusE39696
DAkkS factory calibration for a freely selectable temperature for Memmert Incubators.
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) IF260plus Incubator
Product Type DAkkS Factory Calibration