missing translation for 'onlineSavingsMsg'
Learn More

Groesfjeld Diagnostics deconex™ 11 Universal

Suitable for soaking baths and ultrasonic cleaning equipment. Groesfjeld Diagnostics deconex™ 11 Universal is a mildly alkaline concentrate with activated chlorine for decontaminating lab glassware. It can be universally used in different sectors and is an excellent substitute for chromic acid.

Specifications

Form Concentrated Liquid
Appearance White Emulsion
Composition Alkalis, surfactants, inorganic chlorine carrier
Description Suitable for borosilicate, Duran and quartz glass; porcelain; stainless and chrome nickel steel; polyethylene, polypropylene, Teflon, silicone and natural rubber. Not suitable for aluminum, zinc or tin.
For Use With (Application) Cleaning and decontamination
View More Specs

Description

Description

deconex™ 11 Universal exhibits a wide cleaning spectrum due to its composition. This allows even stubborn contaminants to be removed, such as baked-on fats, oils, waxes, dried blood, proteins, residues of culture media, pen markings, pigments, traces of organic and inorganic materials, and silicones.

  • The surfactants used result in excellent wetting of glassware and can be rinsed without residue
  • The combination of surfactants with the alkalis and activated chlorine removes contaminants and prevents them from being re-deposited on the glassware
  • The easily biodegradable product is pH-buffered and intended for use over a period of several days
  • pH (1% solution in demineralized water): approx. 11.8
  • Density (in concentrate): 1.10g/ml
Specifications

Specifications

Concentrated Liquid
Alkalis, surfactants, inorganic chlorine carrier
Cleaning and decontamination
White Emulsion
Suitable for borosilicate, Duran and quartz glass; porcelain; stainless and chrome nickel steel; polyethylene, polypropylene, Teflon, silicone and natural rubber. Not suitable for aluminum, zinc or tin.
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • deconex 11 UNIVERSAL

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.

Supplemental information
  • MIXTURE LIST-Contains : Tetracalcium phosphate, potassium hydroxide, sodium hypochlorite

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.