missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ deconex 11 UNIVERSAL

Liquid cleaning concentrate for manual cleaning of laboratory glassware. Contains active chlorine. deconex 11 UNIVERSAL is suitable for use in immersion and ultrasonic baths. This cleaner is a full substitute for chromosulphuric acid.

Brand:  Borer Chemie™ 500100.00-F10W


 View more versions of this product

Product Code. 17416492

  • 253.00 EUR / 10kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Concentrate for manual cleaning
  • High yield, typical concentration: 20-50 mL/L
  • Rinses off without leaving residues
  • For consistent and flawless results
  • Contains active chlorine
  • Without EDTA and NTA
  • Technical support by the manufacturer
Specifications

Specifications

Liquid
Alkalis, surfactants, inorganic chlorine carrier
Bottle
10 x 1 kg
10 x 0.9 L
10 kg
∼11.6 (1% Solution)
Concentrate
5°C to 25°C
0.2 gal.
22 lb.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • deconex 11 UNIVERSAL

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.

Supplemental information
  • MIXTURE LIST-Contains : tetrapotassium pyrophosphate, potassiumhydroxide, sodium hypochlorate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.