missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ deconex 12 BASIC

Liquid cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory utensils in immersion and ultrasonic baths. Thanks to high emulsifying and dispersing capacity, deconex 12 BASIC is effective against stubborn residues.

Brand:  Borer Chemie™ 500325.00-K05W


Product Code. 16498313

  • 184.00 EUR / 7kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Concentrate for manual cleaning
  • High yield, typical concentration: 20-50 mL/L
  • Rinses off without leaving residues
  • For consistent and flawless results
  • Extra wide cleaning spectrum
  • High swelling and dispersing capacity
  • Also for sensitive glass such as UV cuvettes
  • Without EDTA and NTA
  • Technical support by the manufacturer
Specifications

Specifications

Liquid
Alkalis, complexing agents, dispersing agents, surfactants
Canister
1 x 7 kg
4.9 L
7 kg
∼11.8 (1% Solution)
Concentrate
5°C to 25°C
1.3 gal.
15.4 lb.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • deconex 12 BASIC

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.,Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : TRIPOTASSIUM ORTHOPHOSPHATE

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.