missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ deconex 21 LAB CLF

For automated cleaning of laboratory glassware and utensils. Thanks to its excellent material compatibility, deconex 21 LAB CLF also cleans sensitive objects made of aluminium. An acid rinsing with a deconex product is recommended.

Brand:  Borer Chemie™ 500925.00-KK5W


 View more versions of this product

Product Code. 15430350

  • 114.48 EUR / 6kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Concentrate for automated cleaning
  • High yield, typical concentration: 5-10 mL/L
  • Excellent material compatibility, also for aluminium
  • Broad cleaning spectrum
  • Can be rinsed off without leaving residues
  • For consistent and flawless results
  • Without chlorine, EDTA, NTA and surfactants
  • Acid rinse with deconex cleaner recommended
  • Technical support by the manufacturer
Specifications

Specifications

Liquid
Alkalis, complexing agents, corrosion inhibitor, sequestering agents
Canister
1 x 6 kg
4.4 L
6 kg
∼11.4 (1% Solution)
Concentrate
5°C to 25°C
1.2 gal.
13.2 lb.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • deconex 21 CLF

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : tetrapotassium pyrophosphate, potassiumsilicate, tripotassium phosphate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers