Learn More
Fisherbrand™ Dedicated Platform
Shop All Fisherbrand Products
Click to view available options
Holds:
12 x 125 mL Flasks
4 Microplates
5 x 500 mL Flasks
8 x 250 mL mL Flasks
Unit Size:
Each
Specifications
Specifications
| Holds | 12 x 125 mL Flasks |
| Product Type | Dedicated Platform |
| Material | Aluminum, Stainless Steel |
| For Use With (Equipment) | Mini Shaking Incubator |
| No. of Positions | 12 |