Learn More
Helios Italquartz Deep and Shallow Crucibles, Transparent
Deep and shallow crucibles constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 63P00012
 View more versions of this product
Product Code. 11784056
  • 87.05 EUR / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Description
  • Transparent
  • Deep and shallow forms available
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
50 cc
37 mm
Quartz
57 mm
Crucible
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products