Learn More
Helios Italquartz Deep and Shallow Crucibles, Transparent
Deep and shallow crucibles constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 63P00002
 View more versions of this product
Product Code. 15858044
  • 63.90 EUR / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Description
  • Transparent
  • Deep and shallow forms available
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
5 cc
19 mm
Quartz
29 mm
Crucible