Learn More
Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators
Product Code. 10041442 Shop All Thermo Scientific Products
Click to view available options
Displacement:
116L/min. (at 60Hz); 97L/min. (at 50Hz)
195L/min. (at 60Hz); 162L/min. (at 50Hz)
283L/min. (at 60Hz); 237L/min. (at 50Hz)
76L/min. (at 60Hz); 62L/min. (at 50Hz)
Voltage:
230 V
Unit Size:
Each
Product Code. 10041442
Brand: Thermo Scientific™ VLP80230
Achieve maximum solute recovery with the Thermo Scientific™Deep Vacuum Oil Pump.
Achieve maximum solute recovery with the Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators, designed to work with all Thermo Scientific™ SpeedVac™ concentrating and freeze drying systems.
Features:
  • Three-position gas ballast control optimizes pump performance to suit all applications
  • Large, sideview sight glass allows for convenient checking of proper oil levels at a glance
All Rotary Vane Oil Pumps come standard with:
  • Muffler (OME190 Oil Mist Eliminator)
  • Two Oil Filters (MF190)
  • Quick Drain Kit
TRUSTED_SUSTAINABILITY
Specifications
Description VLP80; 230V 50Hz
Type Deep Vacuum Oil Pump
Displacement 76L/min. (at 60Hz); 62L/min. (at 50Hz)
For Use With (Equipment) Vacuum Concentrator
Oil Capacity 0.7 L
Ultimate Vacuum Level 1.5 mtorr/1.95 x10³mbar
Vacuum Inlet Fitting 12.7 OD
Width (English) 6 in.
Width (Metric) 160 mm
Height (English) 9 in.
Height (Metric) 230 mm
Depth (English) 17 in.
Depth (Metric) 420 mm
Weight (English) 48 lb.
Weight (Metric) 22 kg
Electrical Requirements 230 V 50 Hz
Voltage 230 V
Frequency 50 Hz