missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deoxyribonucleic Acid Sodium Salt, Fisher BioReagents
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
Packaging:
Amber Glass
Description
Deoxyribonucleic Acid Sodium Salt is used in several biochemical research applications.
Specifications
Specifications
Color | White |
Loss on Drying | 14% max. |
Packaging | Amber Glass |
Quantity | 10 g |
Formula Weight | 6000000 |
Physical Form | Solid |
Chemical Name or Material | Deoxyribonucleic Acid Sodium Salt, From Salmon Sperm |
Safety and Handling
Product Identifier
- Deoxyribonucleic Acid, Sodium Salt
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'