Learn More
KGW Isotherm™ Dewar Insulated Carrying Flasks
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
1 L
2 L
3 L
Cap Lid Type:
Blue-coated Metal Cover
Stainless Steel Cover
missing translation for 'unitSize'
Each
Beschreibung
- Cooling of small samples
- Silver-plated and evacuated
- Flexible applications in the laboratory
- Includes handle made from aluminum
- Includes insulated lid, held by two spring clips
- Available with either blue-coated metal cover or stainless steel cover
Spezifikation
Spezifikation
| Cap Lid Type | Stainless Steel Cover |
| Diameter (Metric) Outer | 160mm |
| For Use With (Application) | Cooling of small samples |
| Height (Metric) Inner | 170mm |
| Height (Metric) | 245mm |
| Includes | Insulated lid, held by two spring clips, handle |
| Capacity (Metric) | 2 L |
| Diameter (Metric) Inner | 138 mm |
| Material | Borosilicate Glass |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur