missing translation for 'onlineSavingsMsg'
Learn More

Diacetone acrylamide, 99%, Thermo Scientific Chemicals

36.68 EUR - 371.85 EUR

Chemical Identifiers

CAS 2873-97-4
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD00008788
InChI Key OMNKZBIFPJNNIO-UHFFFAOYSA-N
Synonym diacetone acrylamide, diacetoneacrylamide, n-1,1-dimethyl-3-oxobutyl acrylamide, 2-propenamide, n-1,1-dimethyl-3-oxobutyl, n-2-methyl-4-oxopentan-2-yl acrylamide, n-2-2-methyl-4-oxopentyl acrylamide, acrylamide, n-1,1-dimethyl-3-oxobutyl, n-1,1-dimethyl-3-oxobutyl-2-propenamide, acrylamide, n,n-diacetonyl, ccris 5898
PubChem CID 17888
IUPAC Name N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide
SMILES CC(=O)CC(C)(C)NC(=O)C=C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488476
View Documents
Thermo Scientific Alfa Aesar
A15940.18
50 g
36.68 EUR
50g
Please to purchase this item. Need a web account? Register with us today!
11498476
View Documents
Thermo Scientific Alfa Aesar
A15940.30
250 g
116.58 EUR
250g
Please to purchase this item. Need a web account? Register with us today!
11478476
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15940.0B
1000 g
371.85 EUR
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.

Solubility
Soluble in water, methanol, benzene and tetrahydrofuran.

Notes
Store in cool place. Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
Specifications

Chemical Identifiers

2873-97-4
169.22
OMNKZBIFPJNNIO-UHFFFAOYSA-N
17888
CC(=O)CC(C)(C)NC(=O)C=C
C9H15NO2
MFCD00008788
diacetone acrylamide, diacetoneacrylamide, n-1,1-dimethyl-3-oxobutyl acrylamide, 2-propenamide, n-1,1-dimethyl-3-oxobutyl, n-2-methyl-4-oxopentan-2-yl acrylamide, n-2-2-methyl-4-oxopentyl acrylamide, acrylamide, n-1,1-dimethyl-3-oxobutyl, n-1,1-dimethyl-3-oxobutyl-2-propenamide, acrylamide, n,n-diacetonyl, ccris 5898
N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide

Specifications

2873-97-4
>110°C (230°F)
MFCD00008788
Hygroscopic
diacetone acrylamide, diacetoneacrylamide, n-1,1-dimethyl-3-oxobutyl acrylamide, 2-propenamide, n-1,1-dimethyl-3-oxobutyl, n-2-methyl-4-oxopentan-2-yl acrylamide, n-2-2-methyl-4-oxopentyl acrylamide, acrylamide, n-1,1-dimethyl-3-oxobutyl, n-1,1-dimethyl-3-oxobutyl-2-propenamide, acrylamide, n,n-diacetonyl, ccris 5898
OMNKZBIFPJNNIO-UHFFFAOYSA-N
N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide
17888
99%
54°C to 56°C
C9H15NO2
1928444
14,2963
Soluble in water,methanol,benzene and tetrahydrofuran.
CC(=O)CC(C)(C)NC(=O)C=C
169.22
169.23
Diacetone acrylamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 220-713-2

missing translation for 'rtecsNumber' : AS3475000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only