missing translation for 'onlineSavingsMsg'
Learn More

Diallyldibutyltin, Thermo Scientific™

Product Code. 11344567
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15336-98-8
Molecular Formula C14H28Sn
Molecular Weight (g/mol) 315.088
MDL Number MFCD00014944
InChI Key LCGVYMRFNOWPGQ-UHFFFAOYSA-N
Synonym diallyldibutyltin, diallyldibutylstannane, dibutylbis prop-2-en-1-yl stannane, diallyl dibutyl stannane, diallyldi-n-butyltin, acmc-1bvvj, dibutyl-bis prop-2-enyl stannane
PubChem CID 84873
IUPAC Name dibutyl-bis(prop-2-enyl)stannane
SMILES CCCC[Sn](CCCC)(CC=C)CC=C

Specifications

Density 1.1002 g/mL
Boiling Point 81°C (0.2 mmHg)
Refractive Index 1.5
Quantity 10 g
UN Number UN2788
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 315.07
Physical Form Liquid
Chemical Name or Material Diallyldibutyltin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'