Learn More
KNF Diaphragm Vacuum Pump N 820.3 FT.40.18
Chemically resistant pump for use with highly aggressive or corrosive gases and vapors.
Brand:  KNF 057512/026366
Product Code. 10748491
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • Self-priming vacuum pump.
  • Chemically resistant.
  • For use with highly aggressive or corrosive gases and vapors
  • Integrated KNF self-drying system ensures that condensate is quickly removed from the pump heads without the vacuum being altered. This significantly reduces process time and preserves the pump heads.
  • The vacuum remains constant in the application.
  • Application: For example, for steam sterilizers or vacuum drying on vacuum drying ovens to significantly accelerate the evacuation time.
  • Pump heads made of PTFE, membrane PTFE-coated, valves FFPM.
  • Two heads.
  • High vapor and condensate compatibility.
  • Gas-tight.
Specifications
Specifications
10 mbar absolute
189 x 341 x 239 mm
Pump head: PTFE, Diaphragm: PTFE-coated, Valves: FFPM
40°C
1 bar
230 V, 50 Hz
Connectors for tube: 10 mm ID
9.6 kg
20 L/min.
230 V