Learn More
Vacuubrand™ Diaphragms and Valves Set For MD 1 and MV 0.5
Brand: Vacuubrand™ 20696824
142.50 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
For MD 1 and MV 0.5 | |
1 Set | |
Diaphragm and Valve Set |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92%
Product Content Correction
Product Title
Vacuubrand™ Diaphragms and Valves Set For MD 1 and MV 0.5 > For use with: For MD 1 and MV 0.5
Spot an opportunity for improvement?Share a Content Correction