missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylvinylsilane, 97%, Thermo Scientific Chemicals

25.22 EUR - 229.14 EUR

Chemical Identifiers

CAS 5507-44-8
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00009067
InChI Key MBGQQKKTDDNCSG-UHFFFAOYSA-N
Synonym diethoxy methyl vinyl silane, methylvinyldiethoxysilane, silane, ethenyldiethoxymethyl, vinylmethyldiethoxysilane, diethoxy methyl vinylsilane, silane, diethoxymethylvinyl, vinyldiethoxymethylsilane, vinylmethyldiathoxysilan, diethoxymethylvinyl silane, diethoxy-methyl-vinyl-silane
PubChem CID 79646
IUPAC Name ethenyl-diethoxy-methylsilane
SMILES CCO[Si](C)(C=C)OCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11349055
SDS View Documents
Thermo Scientific Alfa Aesar
L14224.06
5 g N/A N/A N/A
11359055
SDS View Documents
Thermo Scientific Alfa Aesar
L14224.14
25 g
75.48 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
11369055
SDS View Documents
Thermo Scientific Alfa Aesar
L14224.22
100 g
229.14 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 226-850-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethoxymethylvinylsilane, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.