missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-decylphosphonate, 97%, Thermo Scientific Chemicals

Product Code. 11317856
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16165-68-7
Molecular Formula C14H31O3P
Molecular Weight (g/mol) 278.373
MDL Number MFCD00015593
InChI Key HYQXIVAZQBYBAF-UHFFFAOYSA-N
Synonym diethyl 1-decylphosphonate, diethyl decylphosphonate, decylphosphonic acid diethyl ester, decyldiethoxyphosphino-1-one, acmc-20al9l, diethyl n-decanephosphonate, diethyl 1-decanephosphonate, diethyl 1-decyl phosphonate, decyl-phosphonic acid diethyl ester, n-decylphosphonic acid diethyl ester
PubChem CID 2773697
IUPAC Name 1-diethoxyphosphoryldecane
SMILES CCCCCCCCCCP(=O)(OCC)OCC

Specifications

Boiling Point 128°C to 130°C (0.2 mmHg)
Refractive Index 1.439
Quantity 5 g
Beilstein 1784261
Formula Weight 278.38
Percent Purity 97%
Chemical Name or Material Diethyl 1-decylphosphonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'