Learn More
Diethyl dicarbonate, 97%, Thermo Scientific Chemicals
30.25 EUR - 249.24 EUR
Chemical Identifiers
CAS | 1609-47-8 |
---|---|
Molecular Formula | C6H10O5 |
Molecular Weight (g/mol) | 162.14 |
MDL Number | MFCD00009106 |
InChI Key | FFYPMLJYZAEMQB-UHFFFAOYSA-N |
Synonym | diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid |
PubChem CID | 3051 |
ChEBI | CHEBI:59051 |
IUPAC Name | ethoxycarbonyl ethyl carbonate |
SMILES | CCOC(=O)OC(=O)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499767
|
Thermo Scientific Alfa Aesar
B22753.06 |
5 g |
30.25 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409777
|
Thermo Scientific Alfa Aesar
B22753.14 |
25 g |
98.79 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419777
|
Thermo Scientific Alfa Aesar
B22753.22 |
100 g |
249.24 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ribonuclease inhibitorDiethyl dicarbonate acts as an inhibitor of ryanodine binding to ryanodine/calcium(II) receptor channel. It is useful for specific inactivation of nucleases during isolation of undegraded polynucleotides. Further, it inhibits platelet-activating factor acetyl hydrolase. In addition to this, it is involved in the modification reagent for His and Tyr residues in proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRibonuclease inhibitorDiethyl dicarbonate acts as an inhibitor of ryanodine binding to ryanodine/calcium(II) receptor channel. It is useful for specific inactivation of nucleases during isolation of undegraded polynucleotides. Further, it inhibits platelet-activating factor acetyl hydrolase. In addition to this, it is involved in the modification reagent for His and Tyr residues in proteins.
Solubility
Miscible with chloroform, ethanol, esters, ketones and hydrocarbons. Slightly miscible with water.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids, strong bases and ammonia.
Chemical Identifiers
1609-47-8 | |
162.14 | |
FFYPMLJYZAEMQB-UHFFFAOYSA-N | |
3051 | |
ethoxycarbonyl ethyl carbonate |
C6H10O5 | |
MFCD00009106 | |
diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid | |
CHEBI:59051 | |
CCOC(=O)OC(=O)OCC |
Specifications
1609-47-8 | |
93°C to 94°C (18 mmHg) | |
0.97 | |
1.398 | |
637031 | |
14,7998 | |
Miscible with chloroform,ethanol,esters,ketones and hydrocarbons. Slightly miscible with water. | |
CCOC(=O)OC(=O)OCC | |
162.14 | |
CHEBI:59051 | |
97% | |
Diethyl dicarbonate |
1.101 | |
69°C (156°F) | |
C6H10O5 | |
MFCD00009106 | |
Moisture sensitive | |
diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid | |
FFYPMLJYZAEMQB-UHFFFAOYSA-N | |
ethoxycarbonyl ethyl carbonate | |
3051 | |
162.14 | |
Liquid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H332-H335
missing translation for 'einecsNumber' : 216-542-8
missing translation for 'rtecsNumber' : LQ9350000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Moisture Sensitive
Recommended Storage : Keep cold; Store under Nitrogen
- Diethyl dicarbonate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Diethyl pyrocarbonate
RUO – Research Use Only