missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Yeast Extract, ultra-filtered (UF)

Difco™ Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast.

3650.00 EUR

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
Product Line Difco™
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16219741
View Documents
Gibco™
210934
10 kg
3650.00 EUR
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Difco Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
  • Difco Yeast Extract Low-Dusting (LD) was designed to limit the amount of dust generated compared to other spray-dried yeast extracts
  • It is ultra-filtered and specifically designed for mammalian cell culture applications
  • With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum
  • It has an endotoxin level of less than or equal to 500 EU/g
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Difco™
Mammalian, Insect, Microbial
Dehydrated Culture Media
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.