Learn More
Digi-Sense™ Precalibrated Professional Infrared Thermometers
Product Code. 15830026
Click to view available options
Temperature Range (Metric):
-50°C to +1650°C
Temperature Range (English):
-58°F to +3002°F
Unit Size:
Each
Product Code. 15830026
Brand: Digi-Sense™ undefined
Includes
Type K flexible probe, USB cable, software, tripod, carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Digi-Sense™ Precalibrated Professional infrared Thermometers safely and easily locate hot and cold spots, preventing problems and saving costs
  • Reliable measurements
  • Precalibrated to NIST-traceable standards
  • High distance-to-sight (D/S) ratios handle your long-distance measurements
  • External type K input for contact measurements
  • Dual-laser sighting to pinpoint target areas
  • Protective lens cover with magnetic lock
  • Pistol-grip handle is comfortable and easy to use
  • Measure surface temperatures quickly and accurately
  • USB interface for easy data transmission
Warranty
2 Years
Specifications
For Use With (Application) Noncontact surface temperature measurement
Height (English) 7.5 in.
Height (Metric) 18.9 cm
Includes Type K flexible probe, USB cable, software, tripod, carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Length (Metric) 35.56 cm
Material Plastic
Resolution 0.1° 1° above 1000°
Width (English) 2.25 in.
Product Type Infrared Thermometer
Data Storage Data logging
Max. Temperature (English) 3000°F
Max. Temperature (Metric) 1650°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +3002°F
Temperature Range (Metric) -50°C to +1650°C
Calibrated NIST-traceable
For Use With (Equipment) Type-K thermocouple probes with mini connector
Battery Type 9V
Accuracy ±5.4°F from -58° to 68°F; ±1.0% of reading ± 1.8°F from 68° to 752°F; ±1.5% of reading ± 3.6°F from 752° to 1472°F; ±2.5% of reading from 1472° to 3002°F (±3.0°C from -50° to 20°C; ±1.0% of reading ± 1.0°C from 20° to 400°C; ±1.5% of reading ± 2.0°C from 400° to 800°C; ±2.5% of reading from 800° to 1650°C)
Display Backlit LCD
Width (Metric) 5.7 cm
Scale 30:1 Distance:Target Size Ratio