missing translation for 'onlineSavingsMsg'
Learn More

Dihydrotanshinone I, MedChemExpress

MedChemExpress Dihydrotanshinone I is a natural compound extracted from Salvia miltiorrhiza Bunge which has been widely used for treating cardiovascular diseases. Dihydrotanshinone I exhibits entry-blocking effect for MERS-CoV.

167.40 EUR - 518.40 EUR

Specifications

Chemical Name or Material Dihydrotanshinone I
CAS 87205-99-0
Color Red
Purity Grade Notes Research
Molecular Formula C18H14O3
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16484817
Encompass_Preferred
View Documents
Medchem Express
HY-N0360
50 mg
518.40 EUR
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16483856
Encompass_Preferred
View Documents
Medchem Express
HY-N0360
25 mg
351.00 EUR
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16438958
Encompass_Preferred
View Documents
Medchem Express
HY-N0360
10 mg
167.40 EUR
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dihydrotanshinone I is a natural compound extracted from Salvia miltiorrhiza Bunge which has been widely used for treating cardiovascular diseases. Dihydrotanshinone I exhibits entry-blocking effect for MERS-CoV.
Specifications

Specifications

Dihydrotanshinone I
Red
C18H14O3
O=C(C1=C2C=CC3=C1C=CC=C3C)C(C4=C2OC[C@@H]4C)=O
278.3
Research
COVID-19-anti-virus
87205-99-0
Research
DMSO : 2 mg/mL (7.19 mM; Need ultrasonic)
278.3
98.74%
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.