missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl allylmalonate, 97%, Thermo Scientific Chemicals

Product Code. 11307685
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Notes
Protect from moisture, heat. Incomaptible with oxidizing agents. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40637-56-7
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00014959
InChI Key VZNFVLWVVHHMBG-UHFFFAOYSA-N
Synonym dimethyl allylmalonate, dimethyl 2-allylmalonate, propanedioic acid, 2-propenyl-, dimethyl ester, 1,3-dimethyl 2-prop-2-en-1-yl propanedioate, dimethyl allyl-malonate, acmc-1bnbn, dimethyl 2-allylmalonate #, 2-allyl-propanedioic acid dimethyl ester, 2-prop-2-enylpropanedioic acid dimethyl ester
PubChem CID 574826
IUPAC Name dimethyl 2-prop-2-enylpropanedioate
SMILES COC(=O)C(CC=C)C(=O)OC

Specifications

Melting Point <-80°C
Density 1.074
Boiling Point 207°C
Flash Point 95°C (203°F)
Refractive Index 1.435
Quantity 10 g
Beilstein 1772822
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material Dimethyl allylmalonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'