Learn More
Dimethylglyoxime, 99%, Thermo Scientific Chemicals
61.10 EUR - 731.64 EUR
Chemical Identifiers
CAS | 95-45-4 |
---|---|
Molecular Formula | C4H8N2O2 |
Molecular Weight (g/mol) | 116.12 |
MDL Number | MFCD00002117 |
InChI Key | JGUQDUKBUKFFRO-GGWOSOGESA-N |
Synonym | biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky |
PubChem CID | 5356010 |
IUPAC Name | N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine |
SMILES | C\C(=N/O)\C(\C)=N\O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11495173
|
Thermo Scientific Alfa Aesar
A12810.22 |
100 g |
61.10 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11405183
|
Thermo Scientific Alfa Aesar
A12810.36 |
500 g |
208.04 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415183
|
Thermo Scientific Alfa Aesar
A12810.0E |
2500 g |
731.64 EUR
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
Solubility
Soluble in alcohol, ether and solutions of alkalies. Insoluble in water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
95-45-4 | |
116.12 | |
JGUQDUKBUKFFRO-GGWOSOGESA-N | |
5356010 | |
C\C(=N/O)\C(\C)=N\O |
C4H8N2O2 | |
MFCD00002117 | |
biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine |
Specifications
95-45-4 | |
C4H8N2O2 | |
506731 | |
biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
JGUQDUKBUKFFRO-GGWOSOGESA-N | |
N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine | |
5356010 | |
99% |
∼240°C (decomposition) | |
MFCD00002117 | |
14,3246 | |
Soluble in alcohol,ether and solutions of alkalies. Insoluble in water. | |
C\C(=N/O)\C(\C)=N\O | |
116.12 | |
116.12 | |
Dimethylglyoxime |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P210-P240-P241-P264b-P270-P280-P301+P310-P330-P370+P378q-P501c
H228-H301-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III
missing translation for 'einecsNumber' : 202-420-1
missing translation for 'rtecsNumber' : EK2975000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Dimethylglyoxime
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
- H301-Toxic if swallowed.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : 2,3-Butanedione, dioxime
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.