Learn More
Diphenhydramine hydrochloride, 99%, Thermo Scientific Chemicals
42.61 EUR - 112.56 EUR
Chemical Identifiers
CAS | 147-24-0 |
---|---|
Molecular Formula | C17H22ClNO |
Molecular Weight (g/mol) | 291.82 |
MDL Number | MFCD00012479 |
InChI Key | PCHPORCSPXIHLZ-UHFFFAOYSA-N |
Synonym | diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl |
PubChem CID | 8980 |
ChEBI | CHEBI:4637 |
IUPAC Name | 2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride |
SMILES | [H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10193581
|
Thermo Scientific Acros
350850250 |
25 g | Glass bottle |
42.61 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307331
|
Thermo Scientific Acros
350851000 |
100 g | Glass bottle |
112.56 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
147-24-0 | |
291.82 | |
PCHPORCSPXIHLZ-UHFFFAOYSA-N | |
8980 | |
2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride |
C17H22ClNO | |
MFCD00012479 | |
diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl | |
CHEBI:4637 | |
[H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
147-24-0 | |
White | |
98.5% min. (Argentometry) | |
MFCD00012479 | |
diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl | |
PCHPORCSPXIHLZ-UHFFFAOYSA-N | |
2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride | |
8980 | |
291.82 | |
Adhering Crystals or Crystalline Powder |
168°C to 172°C | |
Authentic | |
C17H22ClNO | |
15, 3339 | |
Solubility in water: 1000g/L. Other solubilities: 500g/L ethanol | |
[H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1 | |
291.82 | |
CHEBI:4637 | |
99% | |
Diphenhydramine hydrochloride, 99% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-687-2
missing translation for 'rtecsNumber' : KR7000000
- Diphenhydramine hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.