Learn More
SCAT™ Disc Sensor Set
Shop All SCAT Europe Products
Click to view available options
Includes:
Disc Sensor Set, Signal Lamp, Empty State
Disc Sensor Set, Signal Lamp, Full State
Signal Lamp EU/UK/US, Magnetic Holder, Cable (3 m), Hook and Loop Fastener (2 m), Empty State
Signal Lamp EU/UK/US, Magnetic Holder, Cable (3 m), Hook and Loop Fastener (2 m), Full State
Signal Lamp, Magnetic Holder, Cable (3 m)
Unit Size:
1 set
1 sheet
Description
- Signal lamp - EU / UK / US, for level control, visual signal (flashing) when the level is reached
- Magnetic holder for individual attachment
- Disc sensor, visual alarm at full state
- 3 m signal cable
- 2 m hook and loop fastener for disc sensor
- Includes power supply: 24V DC EU / UK / US plug
Specifications
Specifications
| Certifications/Compliance | CE |
| Includes | Signal Lamp EU/UK/US, Magnetic Holder, Cable (3 m), Hook and Loop Fastener (2 m), Full State |
| For Use With (Equipment) | Disposal Container |
| Product Type | Signal Lamp |