Learn More
Buerkle™ Disposable Funnel for Powders, LaboPlast™/SteriPlast™
Shop All Bürkle Products
Click to view available options
Color:
Blue
White
Material:
Bio-PE
Polystyrene
Polystyrene, Detectable
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Description
- For easy and reliable transferring
- Wide opening prevents clogging
- Suitable for use in laboratories and clean rooms or for sampling
- Clean room manufactured (clean room class 7)
- Individually packaged and optionally sterilized
- Complies with EU food requirements and FDA regulations
Specifications
Specifications
| Diameter (Metric)Top | 100 mm |
| Diameter (Metric) Stem | 10 mm |
| Material | Bio-PE |
| Color | White |
| For Use With (Application) | Filling of Liquids |
| Sterility | Non-sterile |
| Product Type | Filling Funnel (Powders) |