Learn More
Nexcelom™ Disposable Hemacytometer
Plastic cell counting chamber slides with 2 chambers per slide containing a hemacytometer grid on the slide
282.00 EUR
Specifications
Capacity (Metric) 20 microliters
View More Specs
Products 1
Product Code Brand Total Count Price Quantity & Availability  
Product Code Brand Total Count Price Quantity & Availability  
11542196
Encompass
View Documents
Nexcelom™
CP2-001
25 slides for 50 counts
282.00 EUR
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
Description
Description
  • An enclosed chamber with two ports for sample introduction
  • The chamber contains precisely spaced lines in a grid pattern
  • There are two counting chambers per slide for replicates
  • The counting grid pattern is the Improved Neubauer, as in a common Hemacytometer. The consistent pattern design allows the standard cell counting procedure to be followed.
Specifications
Specifications
20 microliters