Learn More
Kartell™ Technicon™ Beckman™, Konelab™ Polystyrene Sample Cups
Product Code. p-8000300
Click to view available options
Capacity (Metric):
1.5 mL
2 mL
4 mL
Height (Metric):
22.6
24.9 mm
37.9 mm
Unit Size:
Pack of 1000
Product Code. 12039877
Brand: Kartell™ 02508004
Only null left
Add to basket
Add to basket
Manufactured with crystal clear polystyrene for optimum visibility. Kartell™ Technicon™ Beckman™ Konelab™ Sample Cups are used for specimen analysis.
  • A range of crystal clear polystyrene auto analyser cups with conical bases
  • For Technicon™, Beckman™, Konelab™
  • Supplied without caps
  • Graduated
Specifications
Quantity 1000/pk.
Capacity (Metric) 4 mL
Color Clear
Product Type Sample Cup
Material Polystyrene
Disposable Single-use
For Use With (Equipment) Technicon, Beckman and Konelab analyzers
Height (Metric) 37.9 mm