missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Disposal Bag w/Biohazard symbol and print
For hygienic disposal of contaminated laboratory items. Greiner Bio-One Disposal Bag is available with w/Biohazard symbol and print on the bag.
Brand: Greiner Bio-One 646203
This item is not returnable.
View return policy
Description
- Non-sterile
- Polypropylene (PP) suitable for steam autoclaves
- Nominal capacities from 10L to 130L
- For single use only
Specifications
w/Biohazard symbol and print | |
65 L | |
Hygienic disposal of contaminated laboratory items | |
0.05 mm |
Autoclavable | |
Translucent | |
Polypropylene | |
780 x 600mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction