Learn More
IKA™ Distillation Receiver
860.00 EUR - 941.00 EUR
Specifications
For Use With (Application) | For Simultaneous Distillation in 5 Evaporation Flasks |
---|---|
For Use With (Equipment) | For Simultaneous Distillation in 5 Evaporation Flasks |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11875922
|
IKA™
0003740900 |
100 mL |
941.00 EUR
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
11865922
|
IKA™
0003740800 |
50 mL |
860.00 EUR
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
Specifications
For Simultaneous Distillation in 5 Evaporation Flasks | |
For Simultaneous Distillation in 5 Evaporation Flasks |