missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMAP1 Rabbit anti-Human, Mouse, Rat, Clone: 5Z8C8, Novus Biologicals™
Rabbit Monoclonal Antibody
196.00 EUR - 494.00 EUR
Specifications
Antigen | DMAP1 |
---|---|
Clone | 5Z8C8 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331307
|
Bio-Techne
NBP3-15740-20UL |
20 μg |
196.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18387375
|
Bio-Techne
NBP3-15740-100UL |
100 μg |
494.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DMAP1 Monoclonal antibody specifically detects DMAP1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
DMAP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Chromatin Research, DNA Repair, MAP Kinase Signaling | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
55929 | |
IgG | |
Affinity purified |
5Z8C8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DNA methyltransferase 1 associated protein 1, DNA methyltransferase 1-associated protein 1, DNMAP1DNMT1-associated protein 1, DNMT1 associated protein 1, DNMTAP1, EAF2, FLJ11543, KIAA1425DKFZp686L09142, MEAF2, SWC4 | |
A synthetic peptide corresponding to a sequence within amino acids 368-467 of human DMAP1 (Q9NPF5). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title