Learn More
Juchheim Laborgeraete™ Double Bosshead
High quality double socket made of malleable cast iron.
13.05 EUR - 25.50 EUR
Specifications
Material Tempered Cast Iron
View More Specs
Products 3
Product Code Brand Height (Metric) Includes Price Quantity & Availability  
Product Code Brand Height (Metric) Includes Price Quantity & Availability  
10189110
View Documents
Juchheim Laborgeraete™
04820 S
20 mm Safety screws for fixing by hand This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10179110
View Documents
Juchheim Laborgeraete™
04810 S
30 mm Safety screws for fixing by hand
25.50 EUR
Each
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
15818122
View Documents
Juchheim Laborgeraete™
04810
30 mm Lobe screws for mounting by hand
13.05 EUR
Each
Description
Description
  • Durable due to robust malleable cast iron
  • Option of safety screws for fixing by hand or lobe screws for mounting by hand
  • Made in Germany
Specifications
Specifications
Tempered Cast Iron