Learn More
Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns
Separate challenging peptide mapping samples with Thermo Scientific™ Double nanoViper™ PepMap™ Neo UHPLC columns.
Brand: Thermo Scientific™ SNV150150PN
Description
Separate challenging peptide mapping samples with Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC columns. These columns feature flexible connectivity, high reproducibilty, and excellent separation performance. Our PepMap Neo columns are packed to higher pressure and provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to reliably obtain greater sample coverage.
Benefits of PepMap Neo Columns
- Brilliant Connectivity — Double nanoViper™ columns connect with ease.
- Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
- Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
Specifications
Analytical Column, Single nanoViper | |
150 mm | |
2 μm | |
100 Å | |
60°C | |
C18 | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase | |
Peptide Mapping, LC-MS |
150 µm | |
22,000 psi (1500 bar) | |
2 to 8 | |
450 m2/g | |
L1 | |
Single nanoViper | |
15% | |
Yes | |
Single nanoViper PepMap Neo 2 µm C18 150 µm x 150 mm |