missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Burker Counting Chambers

Providing precision measuring to count the number of particles per volume unit of a liquid. Karl Hecht™ Assistent™ Burker Counting Chambers are for counting thrombocytes or erythrocytes for export outside EU only.

Brand:  Karl Hecht™ 44372

Product Code. 10319012

  • 172.00 EUR / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Chamber depth of 0.1mm
  • 9 large squares - surface area 1mm2 each = 9mm2
  • Each large square is divided into 16 groups of squares 0.2 x 0.2mm
  • Central support coated by a rhodium layer to provide sharp contrast and brightly illuminated lines
Specifications

Specifications

Chamber depth 0.1mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.